missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride diethyl etherate, 98+%

Product Code. 11334579
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334579 500 mL 500mL
11324579 100 mL 100mL
11432338 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334579 Supplier Thermo Scientific Alfa Aesar Supplier No. A15275.AP
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

Solubility
Miscible with ether and alcohol.

Notes
Moisture sensitive. Store in cool place. Recommended to use under inet atomosphere. Incompatible with metals, acids, bases, alcohols, alkali metals, oxidizing agents and water. Reacts violently with water.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
Synonym Boron trifluoride etherate; Boron fluoride-ether
PubChem CID 8000
IUPAC Name ethoxyethane;trifluoroborane
SMILES FB(F)F.CCOCC

Specifica

Melting Point -60°C
Density 1.13
Boiling Point 125°C to 126°C
Flash Point 47°C (116°F)
Odor Acrid
Linear Formula BF3€O(CH2CH3)2
Refractive Index 1.344
Quantity 500 mL
UN Number UN2604
Beilstein 3909607
Sensitivity Moisture sensitive
Merck Index 14,1350
Solubility Information Miscible with ether and alcohol.
Formula Weight 141.93
Percent Purity ≥98%
Chemical Name or Material Boron trifluoride diethyl etherate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.