missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride diethyl etherate, 46.5% BF{3} min, packaged under Argon in resealable ChemSeal™ bottles

Code produit. 11358958
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11358958

Brand: Thermo Scientific Alfa Aesar 041796.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol

Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
PubChem CID 8000
SMILES FB(F)F.CCOCC

Specifications

Boiling Point 125°C to 126°C
Melting Point -60°C
Physical Form Liquid
Quantity 100 g
Linear Formula BF3·O(CH2CH3)2
UN Number UN2604
Beilstein 3909607
Flash Point 47°C (116°F)
Merck Index 14,1350
Solubility Information Miscible with ether and alcohol.
IUPAC Name ethoxyethane;trifluoroborane
Formula Weight 141.93
Concentration or Composition (by Analyte or Components) 46.5% BF3 min.
Odor Acrid
Packaging ChemSeal™ bottles
Refractive Index 1.344
Sensitivity Moisture sensitive
Density 1.130 g/mL
Chemical Name or Material Boron trifluoride diethyl etherate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.