Learn More
Boron trifluoride diethyl etherate, 46.5% BF{3} min, packaged under Argon in resealable ChemSeal™ bottles
Description
Boron trifluoride diethyl etherate is used as a Lewis acid catalyst in Mukaiyama aldol addition, alkylation, acetylation, isomerization, dehydrations and condensation reactions. It is involved in the prepattion of polyethers in polymerization reactions. As a catalyst, it is used in the preparation of cyclopentyl- and cycloheptyl[b]indoles and other diborane. It is also used in sensitive neutron detectors in ionization chambers as well as monitoring radiation levels in earth’s atmosphere.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Especificaciones
| Boiling Point | 125°C to 126°C |
| Melting Point | -60°C |
| Physical Form | Liquid |
| Quantity | 500 g |
| Linear Formula | BF3·O(CH2CH3)2 |
| UN Number | UN2604 |
| Beilstein | 3909607 |
| Flash Point | 47°C (116°F) |
| Merck Index | 14,1350 |
| Solubility Information | Miscible with ether and alcohol. |
| Mostrar más |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.