missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™

Product Code. 10133254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133254 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10133254 Supplier Thermo Scientific Acros Supplier No. 428141000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-04-4 | C8H18BF3O | 198.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 593-04-4
Color Brown to Yellow
Density 0.9590g/mL
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 30-35%
Packaging AcroSeal™ Glass Bottle
Molecular Formula C8H18BF3O
Linear Formula BF3O(CH2CH2CH2CH3)2
MDL Number MFCD00013195
Quantity 100 mL
Specific Gravity 0.959
Synonym 1-butoxybutane; trifluoroborane, boron trifluoride dibutyl etherate, boron trifluoride-butyl ether complex, boron, trifluoro 1,1'-oxybis butane-, t-4, boron trifluoride; di-n-butyl ether, 1-butoxybutane-trifluoroborane 1:1, 1-butoxybutane; tris fluoranyl borane, trifluoro 1,1'-oxybis butane boron
InChI Key PVKVBDZRILNPJY-UHFFFAOYSA-N
SMILES FB(F)F.CCCCOCCCC
IUPAC Name 1-butoxybutane;trifluoroborane
Molecular Weight (g/mol) 198.04
PubChem CID 68970
Formula Weight 198.03
Physical Form Liquid
Chemical Name or Material Boron trifluoride dibutyl etherate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride dibutyl etherate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.