missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride acetic acid complex, TRC
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
Specifications
Specifications
| Chemical Name or Material | Boron trifluoride acetic acid complex |
| CAS | 373-61-5 |
| Molecular Formula | BF3.2x(C2H4O2) |
| Quantity | 50 g |
| Synonym | Trifluoroborane acetic acid complex |
| InChI Formula | InChI=1S/C4H6BF2O4.F/c1-3(8)10-5(6,7)11-4(2)9;/h1-2H3;/q;-1/p+1 |
| SMILES | O=C(OB(F)(F)OC(C)=O)C.[F-].[H+] |
| IUPAC Name | Acetic acid-boron trifluoride complex; |
| Molecular Weight (g/mol) | 187.91 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction