missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride - acetic acid complex, ca. 33% BF3

Product Code. 10629014 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
This item is not returnable. View return policy

Product Code. 10629014

Brand: Thermo Scientific Acros 277220025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron trifluoride - acetic acid complex
Name Note ca. 33% BF3
CAS 373-61-5
Melting Point -47°C
Color Brown-Yellow to Yellow
Density 1.3450g/mL
Boiling Point 140°C to 148°C
Flash Point 84°C
Packaging Glass bottle
Molecular Formula C4H8BF3O4
Linear Formula BF3·2CH3COOH
MDL Number MFCD00036359
Quantity 2.5 L
Beilstein 04, 115
Fieser 16,176
Specific Gravity 1.345
Synonym boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4
Solubility Information Solubility in water: reacts
InChI Key OVYKFNOIPQYMRU-UHFFFAOYSA-K
SMILES [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O
IUPAC Name acetic acid;trifluoroborane
Molecular Weight (g/mol) 187.91
PubChem CID 11116786
Formula Weight 187.92
Percent Purity ca. 33% BF3
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride - acetic acid complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.