missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-acetic acid complex,35-36.5% BF3

Product Code. 11400363
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
This item is not returnable. View return policy

Product Code. 11400363

Brand: Thermo Scientific Alfa Aesar A11471.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.

Solubility
It reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Stable, but moisture sensitive. Incompatible with water, moist air, metals, strong oxidizing agents. Store under dry atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-61-5
Molecular Formula C4H8BF3O4
Molecular Weight (g/mol) 187.91
MDL Number MFCD00036359
InChI Key OVYKFNOIPQYMRU-UHFFFAOYSA-K
Synonym boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4
PubChem CID 11116786
IUPAC Name acetic acid;trifluoroborane
SMILES [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O

Specifications

Melting Point -47°C
Density 1.353
pH <1
Boiling Point 143°C to 146°C
Flash Point 83°C (181°F)
Odor Pungent
Refractive Index 1.367
Quantity 250 mL
UN Number UN1742
Beilstein 3686177
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 187.91
Percent Purity 96%
Chemical Name or Material Boron trifluoride-acetic acid complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Boron trifluoride-acetic acid complex, 35-36.5% BF3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Boron trifluoride-acetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.