Learn More
Boron trichloride, 1M soln. in dichloromethane, stab.
Description
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Boron trichloride is used in semiconductor production and in the preparation of other boron compounds. It serves as a reagent in the preparation of various protease inhibitors. It is also used as a catalyst and soldering flux. It is involved in the improvement of aluminum castings by treating with boron trichloride vapors. It is utilized to produce electrical resistors. It is also used as a metal refining agent, especially for aluminum, magnesium, zinc and copper alloys. Further, it is used in the preparation of nanomaterials such as nanofibers and nanotubes. In addition to this, it is used in the field of high energy fuels and rocket propellants.
Solubility
Miscible with dichloromethane, ethanol, carbon tetrachloride, diethyl ether, dimethyl formamide, aromatic solvents, saturated and halogenated hydrocarbon.
Notes
Moisture sensitive. Incompatible with water, oxidizing agents, bases and metals. Store in a cool place.
Specifications
Spécification
| Quantity | 100 mL |
| UN Number | UN2927 |
| Solubility Information | Miscible with dichloromethane,ethanol,carbon tetrachloride,diethyl ether,dimethyl formamide,aromatic solvents,saturated and halogenated hydrocarbon. |
| Formula Weight | 117.17 |
| Concentration or Composition (by Analyte or Components) | 1M soln. |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Density | 1.326 |
| Chemical Name or Material | Boron trichloride, stabilized |
Sécurité et traitement
- Boron trichloride, 1M soln. in dichloromethane
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Dichloromethane, Boron trichloride
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.