missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron tribromide, 99% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
As a catalyst in the manufacturing of diborane, ultra pure boronBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 90°C |
Melting Point | -46°C |
Color | Colorless to Orange |
Physical Form | Liquid |
Quantity | 250 g |
UN Number | UN2692 |
Merck Index | 14,1347 |
Solubility Information | Miscible with ethanol and carbon tetrachloride. |
Formula Weight | 250.52 |
Percent Purity | ≤99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'