missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M solution in methylene chloride, AcroSeal™

Product Code. 10336774 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336774 800 mL AcroSeal™ Glass bottle 800mL
10525334 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10336774 Supplier Thermo Scientific Acros Supplier No. 427108000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 10294-33-4 | BBr3 | 250.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron tribromide
CAS 10294-33-4, 75-09-2
Quantity 800 mL
Molecular Formula BBr3
Linear Formula BBr3
MDL Number MFCD00011312
Merck Index 15,1349
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILES BrB(Br)Br
IUPAC Name tribromoborane
Molecular Weight (g/mol) 250.52
PubChem CID 25134
Formula Weight 250.52
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.467
Density 1.4670g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron tribromide, 1M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H300-Fatal if swallowed.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.