missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 378198000

Code : 29

Additional Details : CAS Number : 10294-33-4 Weight : 0.69520kg Transport : UN number : 3384 Chem class : 6.1 Pack group : I


 View more versions of this product

Product Code. 10422493

  • £253.00 / 800mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
  Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

90.0°C
10294-33-4,142-82-5
BBr3
−1°C
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
ILAHWRKJUDSMFH-UHFFFAOYSA-N
tribromoborane
25134
AcroSeal™ Glass bottle
0.8690g/mL
Boron tribromide
800 mL
MFCD00011312
15,1349
Solubility in water: reacts
BrB(Br)Br
250.52
250.52
0.869
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Boron tribromide, 1M solution in heptane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only