missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M soln. in dichloromethane

Product Code. 11333716
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333716 25 mL 25mL
11343716 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333716 Supplier Thermo Scientific Alfa Aesar Supplier No. L14880.AC
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10294-33-4 | BBr3 | 250.52 g/mol

Boron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via Wurtz-type reductive coupling reaction and micrometer-sized organic molecule-DNA hybrid structures. It is also used in bromination and cyclization reactions to prepare cyclopenta[1,3]cyclopropa[1,2-b]pyrrolizin-8-one. It acts as a liquid boron source used in p-type deposition processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via Wurtz-type reductive coupling reaction and micrometer-sized organic molecule-DNA hybrid structures. It is also used in bromination and cyclization reactions to prepare cyclopenta[1,3]cyclopropa[1,2-b]pyrrolizin-8-one. It acts as a liquid boron source used in p-type deposition processes.

Solubility
Miscible with ethanol and carbon tetrachloride.

Notes
Moisture sensitive. Reacts violently with water. Incompatible with alcohols, alkali metals, aluminum, sodium oxides and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-33-4
Molecular Formula BBr3
Molecular Weight (g/mol) 250.52
MDL Number MFCD00011312
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
PubChem CID 25134
IUPAC Name tribromoborane
SMILES BrB(Br)Br

Specifications

Quantity 25 mL
Refractive Index 1.434
Sensitivity Moisture sensitive
Density 1.467
UN Number UN3390
Merck Index 14,1347
Solubility Information Miscible with ethanol and carbon tetrachloride.
Formula Weight 250.52
Concentration or Composition (by Analyte or Components) 1M soln. in dichloromethane
Chemical Name or Material Boron tribromide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron tribromide, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane, boron tribromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.