missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron, plasma standard solution, Specpure™, B 10,000μg/mL

Brand:  Alfa Aesar™ 014370.AD

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 50mL

 View more versions of this product

Product Code. 11357927

  • £52.50 / 50mL


Estimated Shipment date
from Supplier 25-01-2018
Add to basket

Specifications

Chemical Identifiers

Boron plasma standard solution
B 10000μg/mL
Specpure™
MFCD00134034

Specifications

Boron plasma standard solution
Specpure™
H3BO3 in 1% NH4OH
50mL
B 10000μg/mL
Liquid
MFCD00134034
Videos
SDS