missing translation for 'onlineSavingsMsg'
Learn More

Boron phosphate

Product Code. 11388457
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388457 100 g 100g
11398457 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388457 Supplier Thermo Scientific Alfa Aesar Supplier No. 032562.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13308-51-5 | BO4P | 105.78 g/mol

Boron phosphate is used as a catalyst for dehydration as well as in other organic reactions. It is employed as a source of phosphates involved in the exchange reaction to prepare metal phosphates. It finds application in refractories, ceramics and flame-retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13308-51-5
Molecular Formula BO4P
Molecular Weight (g/mol) 105.78
MDL Number MFCD00011318
InChI Key RKVCQBPDVHFKCU-UHFFFAOYSA-N
Synonym boron phosphate, boron orthophosphate, boron phosphate b po4, 2,4,5-trioxa-1-phospha-3-borabicyclo 1.1.1 pentane 1-oxide, 2,4,5-trioxa-1??-phospha-3-borabicyclo 1.1.1 pentan-1-one, borophosphoric acid, boron iii phosphate, boron phosphate hydrate, boronphosphate b po4, boron phosphate 100g
PubChem CID 83329
IUPAC Name 2,4,5-trioxa-1$l^{5}-phospha-3-borabicyclo[1.1.1]pentane 1-oxide
SMILES B12OP(=O)(O1)O2

Specifications

Melting Point >1400°C
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 105.79
Odor Odorless
Density 2.81
Chemical Name or Material Boron phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.