missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11396178
Click to view available options
Quantity:
32 g
10 x 32 g
This item is not returnable. View return policy
1303-86-2
B2O3
69.62
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
518682
CHEBI:30163
O=BOB=O
This item is not returnable. View return policy
1303-86-2
B2O3
69.62
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
518682
CHEBI:30163
O=BOB=O

For Use in the Electronics Industry As a Liquid Encapsulent For Czochralski-Type Growth of III-V Semiconductor Single Crystals (E.G., Gaas, Gap

In the electronics industry as a liquid encapsulant for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1860°C
Melting Point ∽450°C
Color White
Physical Form Cylinder
Quantity 32 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Soluble in water. Slightly soluble in methanol.
IUPAC Name oxo[(oxoboranyl)oxy]borane
Formula Weight 69.62
Odor Odorless
Sensitivity Hygroscopic
Appearance White cylinder
Density 2.46±0.01 g/cm3 at 20°C
Chemical Name or Material Boron oxide, (H2O 200ppm)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'