missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride

Product Code. p-4824633
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11348568

Brand: Thermo Scientific Alfa Aesar 040607.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10043-11-5 | BN | 24.82 g/mol

Boron nitride is used as polymorph, cosmetics, paints, dental cements, and pencil leads. It is also used in xerographic process and laser printers as a charge leakage barrier layer of the photo drum, fillers, processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranimine
SMILES B=N

Specifications

Color White
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 24.82
Odor Odorless
Density 3.48
Chemical Name or Material Boron nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.