missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, Refractory Brushable Paint, BN 10%, Thermo Scientific™

Product Code. 11339398
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339398 1 kg 1kg
11349398 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339398 Supplier Thermo Scientific Alfa Aesar Supplier No. 043278.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Color White
Physical Form Paste
Quantity 1 kg
IUPAC Name boranimine
Concentration or Composition (by Analyte or Components) BN 10%
Odor Odorless
Chemical Name or Material Boron nitride, Refractory Brushable Paint
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.