missing translation for 'onlineSavingsMsg'
Learn More

Boron carbide, Thermo Scientific Chemicals

Catalog Number 11368488
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12069-32-8
CB4
55.25
MFCD00011520
INAHAJYZKVIDIZ-UHFFFAOYSA-N
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
123279
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
B12B3B4B1C234
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12069-32-8
CB4
55.25
MFCD00011520
INAHAJYZKVIDIZ-UHFFFAOYSA-N
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
123279
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
B12B3B4B1C234

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2350°C
Physical Form Powder
Quantity 50 g
Merck Index 14,1344
Formula Weight 55.26
Odor Odorless
Density 2.52 g/mL
Chemical Name or Material Boron carbide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'