missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol, TRC

Product Code. 30137820
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137820 50 g 50g
30133365 250 g 250g
30009870 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137820 Supplier Toronto Research Chemicals Supplier No. TRCB67554550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-45-9
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1S,2R,4S)-, Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1S-endo)-, Borneol, (1S,2R,4S)-(-)- (8CI), (1S,2R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol, (-)-(1S,4S)-Borneol, (-)-Borneol, (-)-[(1S)-endo]-Borneol, (-)-endo-Borneol, (1S)-(-)-Borneol, (1S)-endo-(-)-Borneol, (1S)-endo-Borneol, (1S,2R,4S)-Borneol, (2R)-(-)-Borneol, Camphyl alcohol, L-Borneol, Linderol, Ngai camphor, endo-(-)-Borneol, endo-(1S)-(-)-Borneol, l-Borneol
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C)[C@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1
Formula Weight 154.1358
Chemical Name or Material (-)-Borneol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.