missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30309252
Change view
Click to view available options
Quantity:
1 mL
missing translation for 'unitSize'
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309252 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30309252 missing translation for 'mfr' Dr. Ehrenstorfer missing translation for 'supplierNo' DREA10662810AL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Borneol
CAS 464-45-9
Analyte or Component Names (-)-Borneol
Shipping Condition Room Temperature
Molecular Formula C10 H18 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1S,2R,4S)-, Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, (1S-endo)-, Borneol, (1S,2R,4S)-(-)- (8CI), (1S,2R,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol, (-)-(1S,4S)-Borneol, (-)-Borneol, (-)-[(1S)-endo]-Borneol, (-)-endo-Borneol, (1S)-(-)-Borneol, (1S)-endo-(-)-Borneol, (1S)-endo-Borneol, (1S,2R,4S)-Borneol, (2R)-(-)-Borneol, Camphyl alcohol, L-Borneol, Linderol, Ngai camphor, endo-(-)-Borneol, endo-(1S)-(-)-Borneol, l-Borneol
InChI Formula InChI=1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1
SMILES CC1(C)[C@H]2CC[C@]1(C)[C@H](O)C2
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 154.25
Quantity 1 mL
Formula Weight 154.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.