missing translation for 'onlineSavingsMsg'
Learn More

Boric Anhydride, Purified, J.T. Baker™

Product Code. p-5391077 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15517894

Brand: Avantor J.T.Baker 117601

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
SMILES O=BOB=O

Specifications

Quantity 500 g
IUPAC Name oxo(oxoboranyloxy)borane
Formula Weight 69.62
Grade Purified
Packaging Poly Bottle
Specific Gravity 1L = 1.85kg
Chemical Name or Material Boric Anhydride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.