missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Reagent Grade, ≥99.5%, Solstice

Product Code. 15697680
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
10 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
10kg
1kg
2.5kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15697680 5 kg Poly Bottle 5kg
15687680 500 g Poly Bottle 500g
15667680 1 kg Poly Bottle 1kg
15677680 2.5 kg Poly Bottle 2.5kg
15657680 10 kg Drum with inliner 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15697680 Supplier Honeywell Research Chemicals Supplier No. B02525KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.6 to 4 (at 20°C)
Color Colorless
Melting Point 160°C
Density 1.510g/cm3 at 20°C
Odor Odorless
Packaging Poly Bottle
Vapor Pressure 2.6mmHg (20°C)
Quantity 5 kg
Linear Formula H3BO3
Physical Form Solid
UN Number NONH for all modes of transport
Formula Weight 61.83g/mol
Percent Purity ≥99.5%
Chemical Name or Material Boric acid
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.