missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%, Solstice

Product Code. 15693280
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
500g
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15693280 6 x 500 g Pack of 6
15663280 500 g 500g
15633280 1 kg 1kg
15643280 2.5 kg 2.5kg
15673280 6 x 1 kg Pack of 6
15683280 6 x 2.5 kg Pack of 6
15653280 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15693280 Supplier Honeywell-Fluka Supplier No. 311466X500G
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Vapor Pressure 2.6mmHg (20°C)
Quantity 6 x 500 g
Linear Formula H3BO3
UN Number NONH for all modes of transport
Formula Weight 61.83
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Boric Acid, Reag. Ph. Eur. reag. ISO
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.