Learn More
Boric acid, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
£61.20 - £436.00
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11320879
|
Thermo Scientific Alfa Aesar
010659.14 |
25 g |
£61.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330879
|
Thermo Scientific Alfa Aesar
010659.22 |
100 g |
£182.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340879
|
Thermo Scientific Alfa Aesar
010659.36 |
500 g |
£436.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a primary or adjunct buffer in swimming pools, flame retardant, neutron absorber in nuclear power plants, colorant to make flame green, preservative in urine samples, welding flux in combination with borax, or precursor to other chemical compounds. It is used to prevent/remove existing wet and dry rot in timbers, treat wood against fungal/insect attack, prevent athlete’s foot, lubricate carom boards, and reduce surface oxidation on metals at high temperatures. It can be used to produce lubricant for ceramic or metal surfaces, textile fiberglass, induction furnace linings and ceramics, Silly Putty, and glass LCD flat panel displays. It can prevent the high temperature reaction of aluminum with nitrates in pyrotechniques, and neutralize active hydrofluoric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
10043-35-3 | |
1.43515 | |
99.9995% (metals basis) | |
H3BO3 | |
MFCD00011337 | |
14,1336 | |
Soluble in water- solubility increases rapidly with temperature. Soluble in methanol and ethanol. Very slightly soluble in acetone. | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
99.9995% | |
(metals basis) |
∽185°C (decomposition) | |
None | |
BH3O3 | |
Powder | |
1697939 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Puratronic™ | |
Boric acid |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P281-P308+P313-P501c
H360FD
missing translation for 'einecsNumber' : 233-139-2
missing translation for 'rtecsNumber' : ED4550000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only