Learn More
Boric Acid, MP Biomedicals™
Shop All MP Biomedicals ProductsDescription
Boric acid has a single boron; its salt forms complex equilibria in solution, with the ion B4O7-2 being the predominant species. Manufacturing and industrial uses include: for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems; in nickeling baths; cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel. It is also used as an insecticide for cockroaches and black carpet beetles.
Specifications
Specifications
| Quantity | 1 kg |
| Physical Form | White Powder |
| Solubility Information | Soluble in water; insoluble in alcohol; freely soluble in boiling water. |
| Formula Weight | 61.84 |
| Percent Purity | ≥99.5% |
| Grade | ACS Reagent |
Safety and Handling
- Boric Acid, ACS
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Boric Acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.