missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Application include:
- As an astringent, antiseptic, antibacterial and antifungal agent in research labs
- In buffers and for calibration of polyacrylamide gel columns for the separation of oligonucleotides by capillary electrophoresis
- Weatherproofing wood and fireproofing fabrics
- Manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems
- As a preservative and in nickeling baths
- Cosmetics, printing, dyeing, painting and photography
- For impregnating wicks, in electric condensers and for hardening steel
- As an insecticide for cockroaches and black carpet beetles
Specifications
Specifications
| CAS | 10043-35-3 |
| Quantity | 25 g |
| Molecular Formula | BH3O3 |
| Physical Form | White Powder |
| MDL Number | MFCD00011337 |
| Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
| Solubility Information | Soluble in water; insoluble in alcohol; freely soluble in boiling water. |
| InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
| SMILES | OB(O)O |
| IUPAC Name | boric acid |
| Show More |
Safety and Handling
Product Identifier
- Boric Acid, ACS
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Boric Acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?