missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Granular, Multi-Compendial, J.T. Baker™

Product Code. 15507774 Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 15507774

Brand: Avantor J.T.Baker 009205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 10043-35-3
Packaging Poly Bottle
Specific Gravity 1L = 1.43kg
Quantity 2.5 kg
Molecular Formula BH3O3
Physical Form Granular
MDL Number MFCD00011337
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
PubChem CID 7628
ChEBI CHEBI:33118
Formula Weight 61.83
Grade Multi-Compendial
Chemical Name or Material Boric Acid
Show More Show Less
compliance-icons
Product Identifier
  • ortho-Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ortho-Boric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.