missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Solstice

Product Code. 16476349 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Grade:
ACS
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Grade unitSize
16476349 250 g Plastic Bottle ACS 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476349 Supplier Honeywell-Fluka Supplier No. 31146250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%

  • Leading the industry for over 65 years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

pH 3.6 to 4.0 (20°C)
Color Colorless
CAS 10043-35-3
Melting Point 171°C
Packaging Plastic Bottle
Quantity 250 g
Molecular Formula BH3O3
MDL Number MFCD00011337
Synonym Acidum Boricum
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
Percent Purity 99.8%
Grade ACS
Chemical Name or Material Boric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.