Learn More
Boric acid, 99+%, Thermo Scientific Chemicals
£19.80 - £220.00
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484253
|
Thermo Scientific Alfa Aesar
A10896.36 |
500 g |
£19.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494253
|
Thermo Scientific Alfa Aesar
A10896.0E |
2500 g |
£61.50
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404263
|
Thermo Scientific Alfa Aesar
A10896.0C |
10,000 g |
£220.00
10000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
Solubility
Soluble in water and alcohols. Slightly soluble in acetone and pyridine.
Notes
Store in cool place. Moisture sensitive. Incompatible with potassium and acid anhydrides.
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
3.8 to 4.8 | |
∼185°C (decomposition) | |
Odorless | |
BH3O3 | |
MFCD00011337 | |
14,1336 | |
Soluble in water and alcohols. Slightly soluble in acetone and pyridine. | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
≥99% |
10043-35-3 | |
1.435 | |
None | |
H3BO3 | |
1697939 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Boric acid |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P260-P281-P308+P313-P501c
H360FD
missing translation for 'einecsNumber' : 233-139-2
missing translation for 'rtecsNumber' : ED4550000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only