missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, 99+%

Product Code. 11484253
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484253 500 g 500g
11494253 2500 g 2500g
11404263 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484253 Supplier Thermo Scientific Alfa Aesar Supplier No. A10896.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol

Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.

Solubility
Soluble in water and alcohols. Slightly soluble in acetone and pyridine.

Notes
Store in cool place. Moisture sensitive. Incompatible with potassium and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Melting Point ∼185°C (decomposition)
Density 1.435
Odor Odorless
Flash Point None
Quantity 500 g
Linear Formula H3BO3
Beilstein 1697939
Merck Index 14,1336
Solubility Information Soluble in water and alcohols. Slightly soluble in acetone and pyridine.
Formula Weight 61.83
Percent Purity ≥99%
Chemical Name or Material Boric acid
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: boric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.