missing translation for 'onlineSavingsMsg'
Learn More

Melford Boric Acid

Product Code. 30018078
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018078 5 kg 5kg
30018077 500 g 500g
30018076 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018078 Supplier Melford Supplier No. B320505000.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For use in preparing electrophoresis buffers.

For use in preparing electrophoresis buffers.

Specifications

Chemical Name or Material Boric Acid
CAS 10043-35-3
Quantity 5 kg
Molecular Formula H3BO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.