missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, 2% Solution + Indicator, Chem-Lab

Catalog Number 11909182
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for Kjeldahl nitrogen determination and AFNOR determination in water. Boric Acid, 2% Solution + Indicator, Chem-Lab consists of 20g H3BO3 + indicator per 1L water. It has a pH value of 3.90–4.30.

Boric Acid, 2% Solution + Indicator is used in the analysis and quantitative determination of organic nitrogen.
pH 3.90–4.30
Color Purple to Red
CAS 10043-35-3, 7732-18-5
Concentration 20g H3BO3 + indicator per 1L H2O
Density 1.00g/mL
Odor Odorless
Quantity 5 L
Molecular Formula BH3O3
Physical Form Liquid
MDL Number MFCD00011337
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
Solubility Information Water
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES OB(O)O
IUPAC Name boric acid
Molecular Weight (g/mol) 61.83
PubChem CID 7628
ChEBI CHEBI:33118
Formula Weight 61.83g/mol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'