Learn More
BORG5/CDC42EP1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27904-1-AP-150UL
Description
The BORG5/CDC42EP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BORG5/CDC42EP1. This antibody recognizes human antigen. The BORG5/CDC42EP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BORG5/CDC42EP1 | |
| 700 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Binder of Rho GTPases 5, BORG5, Cdc42 effector protein 1, CDC42EP1, CEP1, MSE55, Serum protein MSE55 | |
| 150 μL | |
| 11135 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |