missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ borer 25 ORGANACID

Product Code. 17425112
Click to view available options
Quantity:
1 x 5 kg
1 x 10 kg
1 x 25 kg
Unit Size:
10kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 17425112

Brand: Borer Chemie™ 502100.15KK5R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleaner and neutralizer for laboratory washers. Shortens rinsing phases and reduces water consumption. borer 25 ORGANACID is phosphate-free and suitable for environmental and water laboratories. Pre-rinsing agent against acid-soluble residues.

  • Neutralizer and cleaner for automated use
  • Ecological solution without phosphates and surfactants
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Typical application concentration: 3 mL/L
  • Excellent material compatibility, also for aluminium
  • Can be rinsed off without leaving residues
  • Also as pre-rinsing agent against acid-soluble deposits
  • Technical support by the manufacturer

Specifications

Form Liquid
pH ∼2.6 (1% Solution)
Composition Organic acid
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.1 gal.
Volume (Metric) 4.3 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 25 ORGANACID
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : citric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.