missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ borer 22 LIQ-x

Product Code. p-7227845
Click to view available options
Quantity:
1 x 6 kg
1 x 12 kg
1 x 30 kg
Unit Size:
12kg
30kg
6kg
This item is not returnable. View return policy

Product Code. 17465122

Brand: Borer Chemie™ 519200.15KK5W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Universal use for automated cleaning of laboratory glassware and utensils. Without phosphates, surfactants and chlorine. borer 22 LIQ-x is an ecological solution with an excellent performance. Acidic rinse with a borer product recommended.

  • Concentrate for universal automated cleaning
  • Ecological solution without phosphates and surfactants
  • Without chlorine, EDTA or NTA
  • High yield, typical concentration: 5-10 mL/L
  • Broad cleaning spectrum
  • High performance thanks to high dispersing capacity
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Acid rinse with a borer cleaner recommended
  • Technical support by the manufacturer

Specifications

Form Liquid
pH ∼12.2 (1% Solution)
Composition Alkalis, complexing agents, sequestering agents
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 6 kg
Volume (English) 1.3 gal.
Volume (Metric) 4.9 L
Weight (English) 13.2 lb.
Weight (Metric) 6 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 22 LIQ-x
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.