missing translation for 'onlineSavingsMsg'
Learn More

BMP-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 19 publications

Brand:  Novus Biologicals NBP1-19751

 View more versions of this product

Product Code. 18797923

  • £433.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BMP-2 Polyclonal specifically detects BMP-2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMP-2
Polyclonal
Unconjugated
PBS (pH 7.4) with 0.05% Sodium Azide
BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2
Rabbit
44 kDa
0.1 mL
Embryonic Stem Cell Markers, Stem Cell Signaling Pathway
650
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 0.5 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30700748), Immunohistochemistry-Paraffin 1:50-1:200
P12643, P21274
BMP2
A synthetic peptide made to an internal region of human BMP2 (within residues 250-350) [Swiss-Prot# P12643]
Affinity Purified
RUO
Primary
Human, Mouse, Rat, Canine
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only