missing translation for 'onlineSavingsMsg'
Learn More

Borealin Antibody, DyLight 594, Novus Biologicals™

Product Code. 18427573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427573 Supplier Novus Biologicals Supplier No. NBP177330DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Borealin Polyclonal antibody specifically detects Borealin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borealin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, dasra-B, DasraB, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of the human CDCA8. The immunogen is located within the first 50 amino acids of CDCA8. Amino Acid Squence: KDFDREVEIRIKQIESDR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55143
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.