missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bordetella pertussis Toxin Monoclonal Antibody (1280/204)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183016
This item is not returnable.
View return policy
Description
Mouse anti Bordetella Pertussis Toxin antibody, clone 1280/204 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined.
B. pertussis has the ability to inhibit the function of the host's immune system. The toxin, known as pertussis toxin (or PTx), inhibits G protein coupling that regulates an adenylate cyclase-mediated conversion of ATP to cyclic AMP. The end result is phagocytes convert too much ATP to cyclic AMP, which can cause disturbances in cellular signaling mechanisms, and prevent phagocytes from correctly responding to an infection.
Specifications
| Bordetella pertussis Toxin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 1280/204 | |
| Unconjugated | |
| B. pertussis; PTx | |
| Native protein. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction