missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Borate-buffered saline (5X)

Product Code. 15413659
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. Quantity unitSize
15413659 250 mL 250mL
15423659 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 15413659

Brand: Thermo Scientific™ J60979.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. Borate buffer solutions is used in protein modification procedures requiring amine-free buffer at alkaline pH.

Solubility
Soluble in water 0.1 M at 20°C.

Notes
125mM sodium borate, 750mM sodium chloride, pH 7.0
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Borate-buffered Saline
Concentration 5X
Physical Form Liquid
Quantity 250 mL
Molecular Formula B4Na2O7
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular Weight (g/mol) 201.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.