missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.5

Product Code. 15454579
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454579 500 mL 500mL
15444579 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454579 Supplier Thermo Scientific Chemicals Supplier No. J62154.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

Solubility
Soluble in water.

Notes
Store at 25°C. This product is relatively stable under this condition. Best result obtained with use within 6-8 months of purchase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 9.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water.
Chemical Name or Material Borate buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.