missing translation for 'onlineSavingsMsg'
Learn More

Borane-trimethylamine complex, 97%

Product Code. 10577932 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10577932 100 g Glass bottle 100g
10542384 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10577932 Supplier Thermo Scientific Acros Supplier No. 402741000
  • ONLINE EXCLUSIVE PRICE
    £244.00
    £219.65 / 100g
    Save £24.35
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-22-9 | C3H12BN | 72.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-22-9
Molecular Formula C3H12BN
Molecular Weight (g/mol) 72.95
MDL Number MFCD00012420
InChI Key WVMHLYQJPRXKLC-UHFFFAOYSA-N
Synonym Trimethylamine borane
IUPAC Name trimethylamine borane
SMILES B.CN(C)C

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 172.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)3N·BH3
Quantity 100 g
Fieser 01,1229; 05,708; 12,65
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ethanols,ethyl ether
Formula Weight 72.95
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Borane-trimethylamine complex, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.