missing translation for 'onlineSavingsMsg'
Learn More

Borane-trimethylamine complex, 97%

Product Code. 11446830
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446830 5 g 5g
11456830 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446830 Supplier Thermo Scientific Alfa Aesar Supplier No. L14994.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-22-9 | C3H12BN | 72.95 g/mol

Reducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.

Solubility
Decomposes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-22-9
Molecular Formula C3H12BN
Molecular Weight (g/mol) 72.95
MDL Number MFCD00012420
InChI Key WVMHLYQJPRXKLC-UHFFFAOYSA-N
IUPAC Name trimethylamine borane
SMILES B.CN(C)C

Specifications

Melting Point 93°C to 95°C
Boiling Point 172°C
Flash Point 64°C (147°F)
Quantity 5 g
UN Number UN1325
Beilstein 3905060
Formula Weight 72.95
Percent Purity 97%
Chemical Name or Material Borane-trimethylamine complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.