missing translation for 'onlineSavingsMsg'
Learn More

Borane-triethylamine complex, 96%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 11320248
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11320248 5 g 5g
11330248 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320248 Supplier Thermo Scientific Alfa Aesar Supplier No. 042962.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a reducing agent and in hydroboration reactions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1722-26-5
Molecular Formula C6H15N·BH3
MDL Number MFCD00012423

Specifications

Melting Point -4°C
Color Colorless to Yellow
Density 0.777 g/mL
Boiling Point 97°C (12 mmHg)
Flash Point −6°C (21°F)
Odor Fish-like
Packaging ChemSeal™ bottles
Linear Formula (C2H5)3N·BH3
Quantity 5 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Miscible with water,methanol,diethyl ether,terahydro furan,toluene and dichloromethane. Immiscible with hexane.
Formula Weight 115.03
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Borane-triethylamine complex
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.