missing translation for 'onlineSavingsMsg'
Learn More

Borane-tetrahydrofuran complex, 1M soln. in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15471887
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15471887

Brand: Thermo Scientific Alfa Aesar 043291.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14044-65-6
Molecular Formula C4H11BO
Molecular Weight (g/mol) 85.94
MDL Number MFCD00012429
InChI Key RMCYTHFAWCWRFA-UHFFFAOYSA-N
Synonym borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
PubChem CID 11062302
IUPAC Name boron;oxolane
SMILES B.C1CCOC1

Specifications

Color Colorless
Density 0.878 g/mL
Flash Point -21°C (-8°F)
Packaging ChemSeal™ bottles
Linear Formula BH3/(CH2)4O
Quantity 500 mL
UN Number UN3148
Beilstein 3668402
Sensitivity Air and moisture sensitive
Solubility Information Miscible and reacts violently with water.
Formula Weight 85.94
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Physical Form Liquid
Chemical Name or Material Borane-tetrahydrofuran complex
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.