missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%

Product Code. 10762411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10762411 25 mL 25mL
10723013 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762411 Supplier Thermo Scientific Acros Supplier No. 177010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-51-0 | C5H8BN | 92.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridineborane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Melting Point 10.0°C to 11.0°C
Density 0.9200g/mL
Flash Point 73°C
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.5300 to 1.5320
Quantity 25 mL
Fieser 08,50; 09,59; 12,65
Specific Gravity 0.92
Solubility Information Solubility in water: reacts slowly with water. Other solubilities: soluble in methanol,ethanol
Viscosity 2.5 mm2/s (22°C)
Formula Weight 92.94
Percent Purity 95%
Chemical Name or Material Borane-pyridine complex
Show More Show Less
compliance-icons
Product Identifier
  • Borane-pyridine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H310-Fatal in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.