missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%

Product Code. 11436790
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436790 50 g 50g
11426790 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436790 Supplier Thermo Scientific Alfa Aesar Supplier No. L13178.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-51-0 | C5H8BN | 92.94 g/mol

Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridine-borane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 0.92
Boiling Point 65°C (1 mmHg)
Flash Point 73°C (163°F)
Odor Unpleasant
Refractive Index 1.532
Quantity 50 g
UN Number UN2810
Beilstein 3674913
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes slowly in water.
Formula Weight 92.93
Percent Purity 95%
Chemical Name or Material Borane-pyridine complex
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.