missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%, (ca. 9.4M), AcroSeal™

Product Code. 10041544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041544 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041544 Supplier Thermo Scientific Acros Supplier No. 427131000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-51-0 | C5H8BN | 92.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-51-0
Melting Point 10.0°C to 11.0°C
Color Amber to Colorless
Density 0.9200g/mL
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H8BN
Refractive Index 1.5300 to 1.5320
MDL Number MFCD00012435
Quantity 100 mL
Specific Gravity 0.92
Synonym Pyridineborane
Solubility Information Solubility in water: reacts slowly with water. Other solubilities: soluble in methanol,ethanol
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
SMILES [BH3-][N+]1=CC=CC=C1
IUPAC Name (pyridin-1-ium-1-yl)boranuide
Molecular Weight (g/mol) 92.94
Viscosity 2.5 mm2/s (22°C)
Formula Weight 92.94
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Borane-pyridine complex, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Borane-pyridine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H310-Fatal in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.