missing translation for 'onlineSavingsMsg'
Learn More

Borane-N,N-diethylaniline complex, 97%

Product Code. 10215900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
AcroSeal™ Glass bottle
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215900 25 g Glass bottle 25g
10702801 100 g AcroSeal™ Glass bottle 100g
10414605 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215900 Supplier Thermo Scientific Acros Supplier No. 176770250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13289-97-9 | C10H15BN | 160.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13289-97-9 , 91-66-7
Molecular Formula C10H15BN
Molecular Weight (g/mol) 160.05
MDL Number MFCD00013187
InChI Key KHYAFFAGZNCWPT-UHFFFAOYSA-N
Synonym n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
PubChem CID 6335292
IUPAC Name boron;N,N-diethylaniline
SMILES [B].CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -30°C to 27°C
Color Colorless to Amber
Density 0.9200g/mL
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5N(C2H5)2·BH3
Refractive Index 1.5245 to 1.5265
Quantity 25 g
Specific Gravity 0.92
Solubility Information Solubility in water: may decompose
Formula Weight 163.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Borane-N, N-diethylaniline complex
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Borane-N,N-diethylaniline complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.