missing translation for 'onlineSavingsMsg'
Learn More

Borane-morpholine complex, 97%

Product Code. 11416840
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416840 25 g 25g
11406840 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416840 Supplier Thermo Scientific Alfa Aesar Supplier No. L15110.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4856-95-5 | C4H12BNO | 100.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4856-95-5
Molecular Formula C4H12BNO
Molecular Weight (g/mol) 100.96
MDL Number MFCD00066736
InChI Key IFVAHVOGOBRFSP-UHFFFAOYSA-N
Synonym morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine
PubChem CID 6327109
IUPAC Name boron;morpholine
SMILES B.C1COCCN1

Specifications

Melting Point 93°C to 95°C
Odor Amine-like
Quantity 25 g
UN Number UN2925
Formula Weight 100.96
Percent Purity 97%
Chemical Name or Material Borane-morpholine complex

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.