missing translation for 'onlineSavingsMsg'
Learn More

Borane-diphenylphosphine Complex, 94%

Product Code. 10330933 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330933 1 g Glass bottle 1g
10390973 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10330933 Supplier Thermo Scientific Acros Supplier No. 380750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41593-58-2 | C12H14BP | 200.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41593-58-2
Molecular Formula C12H14BP
Molecular Weight (g/mol) 200.03
MDL Number MFCD00274254
InChI Key XUFGOLYVDOBYLK-UHFFFAOYSA-N
Synonym borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne
PubChem CID 11095625
IUPAC Name boron;diphenylphosphane
SMILES B.P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 47.0°C to 50.0°C
Color White
Elemental Analysis 92.0% min. ex C% min.
Boiling Point 148.0°C to 150.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: decomposes
Formula Weight 200.03
Percent Purity 94%
Physical Form Crystals or Powder
Chemical Name or Material Borane-diphenylphosphine complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Borane-diphenylphosphine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.